Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J2 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502454S


Description
POLR2J2 Monoclonal antibody specifically detects POLR2J2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
POLR2J2 | |
Monoclonal | |
0.72 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WWB5 | |
Mouse | |
Affinity chromatography | |
RUO | |
120379 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3G4 | |
Unconjugated | |
PIH1D2 | |
DNA directed RNA polymerase II polypeptide J-related; DNA-directed RNA polymerase II subunit J2; DNA-directed RNA polymerase II subunit RPB11-b1; HRPB11B; POLR2J2; polymerase (RNA) II (DNA directed) polypeptide J2; polymerase (RNA) II subunit J2; RNA polymerase II subunit B11-b1; RNA polymerase II subunit J2; RPB11b1 | |
Full length human recombinant protein of human PIH1D2 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction