Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2J2 Monoclonal Antibody (OTI6E9), TrueMAB™, OriGene
SDP

Catalog No. 502541537
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1537 30 μL
50-167-3510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1537 Supplier OriGene Supplier No. TA502455S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POLR2J2 Monoclonal antibody specifically detects POLR2J2 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen POLR2J2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6E9
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene POLR2J2
Gene Accession No. Q9GZM3
Gene Alias DNA directed RNA polymerase II polypeptide J-related; DNA-directed RNA polymerase II subunit J2; DNA-directed RNA polymerase II subunit RPB11-b1; HRPB11B; POLR2J2; polymerase (RNA) II (DNA directed) polypeptide J2; polymerase (RNA) II subunit J2; RNA polymerase II subunit B11-b1; RNA polymerase II subunit J2; RPB11b1
Host Species Mouse
Immunogen Full length human recombinant protein of human POLR2J2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246721
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.