Learn More
Invitrogen™ POLR2L Recombinant Rabbit Monoclonal Antibody (3C6H5)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | POLR2L |
---|---|
Clone | 3C6H5 |
Concentration | 1.14 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: MIIPVRCFTC GKIVGNKWEA YLGLLQAEYT EGDALDALGL KRYCCRRMLL AHVDLIEKLL NYAPLEK.
This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains four conserved cysteines characteristic of an atypical zinc-binding domain. Like its counterpart in yeast, this subunit may be shared by the other two DNA-directed RNA polymerases.Specifications
POLR2L | |
1.14 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P62875, P62876 | |
502374, 5441, 66491 | |
A synthetic peptide corresponding to a sequence within amino acids 1-67 of human POLR2L (NP_066951.1). | |
Antibody |
3C6H5 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
POLR2L | |
2510029B14Rik; DNA-directed RNA polymerase III subunit L; DNA-directed RNA polymerases I, II, and III subunit RPABC5; hRPB7.6; hsRPB10b; Polr2l; polymerase (RNA) II (DNA directed) polypeptide L; polymerase (RNA) II (DNA directed) polypeptide L, 7.6kDa; polymerase (RNA) II subunit L; RBP10; RNA polymerase II 7.6 kDa subunit; RNA polymerase II subunit L; RNA polymerases I, II, and III subunit ABC5; RPABC5; RPB10; RPB10 homolog; RPB10beta; RPB7.6 | |
POLR2L | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.