Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3B Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306317DL594
Description
POLR3B Polyclonal specifically detects POLR3B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
POLR3B | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C128, DKFZp686B10117, DNA-directed RNA polymerase III 127.6 kDa polypeptide, DNA-directed RNA polymerase III subunit B, DNA-directed RNA polymerase III subunit RPC2, EC 2.7.7.6, FLJ10388, polymerase (RNA) III (DNA directed) polypeptide B, RNA polymerase III subunit C2, RPC2 | |
Produced in rabbits immunized with E. coli-derived Human POLR3B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55703 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction