Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543229 Supplier Invitrogen™ Supplier No. PA543229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IEKVMISSNA EDAFLIKMLL RQTRRPEIGD KFSSRHGQKG VCGLIVPQED Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 100%.

POLR3B (RNA Polymerase III Subunit B) is a Protein Coding gene. Diseases associated with POLR3B include Leukodystrophy, Hypomyelinating, 8, With Or Without Oligodontia And/Or Hypogonadotropic Hypogonadism and Cerebellar Hypoplasia With Endosteal Sclerosis. Among its related pathways are RNA Polymerase II Transcription Initiation And Promoter Clearance and ATP/ITP metabolism. Gene Ontology (GO) annotations related to this gene include DNA-directed 5'-3' RNA polymerase activity and ribonucleoside binding. An important paralog of this gene is POLR2B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POLR3B
Gene Accession No. Q9NW08
Gene Alias 2700078H01Rik; A330032P03Rik; C128; C85372; DNA-directed RNA polymerase III 127.6 kDa polypeptide; DNA-directed RNA polymerase III B; DNA-directed RNA polymerase III subunit B; DNA-directed RNA polymerase III subunit RPC2; FLJ10388; HLD8; INMAP; interphase nucleus and mitotic apparatus associated protein; POLR3B; polymerase (RNA) III (DNA directed) polypeptide B; polymerase (RNA) III subunit B; RGD1565311; RNA polymerase III subunit B; RNA polymerase III subunit C2; RNA polymerase III subunit RPC2 homolog; RPC2; si:dkey-103i16.3; slim jim; slj; wu:fc20h06
Gene Symbols POLR3B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human POLR3B (aa 866-915).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55703
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.