Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3C Polyclonal Antibody

POLR3C Polyclonal Antibody
Supplier: Thermo Scientific PA5110939
Description
POLR3C Polyclonal Antibody for Western Blot, ICC/IF

Specifications
POLR3C | |
Polyclonal | |
Unconjugated | |
POLR3C | |
4933407E01Rik; DNA-directed III 62 kDa polypeptide; DNA-directed RNA polymerase III subunit C; DNA-directed RNA polymerase III subunit RPC3; hypothetical protein LOC507314; POLR3C; polymerase (RNA) III (DNA directed) polypeptide C; polymerase (RNA) III (DNA directed) polypeptide C (62kD); polymerase (RNA) III subunit C; RNA polymerase III 62 kDa subunit; RNA polymerase III subunit C; RNA polymerase III subunit C3; RPC3; RPC62 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10623 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUI4 | |
POLR3C | |
Recombinant Protein corresonding to Human POLR3C. Recombinant protein control fragment (Product #RP-94512). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction