Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR3E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559585 Supplier Invitrogen™ Supplier No. PA559585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVMWKFTQSR WVVRKEVATV TKLCAEDVKD FLEHMAVVRI NKGWEFILPY DGEFIKKHPD VVQRQHMLWT GIQAKLEKVY NLVKETMPKK PDAQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. Essential for efficient transcription from both the type 2 VAI and type 3 U6 RNA polymerase III promoters. Plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. Acts as nuclear and cytosolic DNA sensor involved in innate immune response. Can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3E
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR3E
Gene Accession No. Q9NVU0
Gene Alias DNA-directed RNA polymerase III 80 kDa polypeptide; DNA-directed RNA polymerase III subunit RPC5; KIAA1452; POLR3E; polymerase (RNA) III (DNA directed) polypeptide E; polymerase (RNA) III (DNA directed) polypeptide E (80kD); polymerase (RNA) III subunit E; RGD1308086; RNA polymerase III 80 kDa subunit RPC5; RNA polymerase III polypeptide E; RNA polymerase III subunit 5; RNA polymerase III subunit C5; RNA polymerase III subunit E; RPC5; Sex-lethal interactor homolog; Sin; Sxl interactor
Gene Symbols POLR3E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR3E. Recombinant protein control fragment (Product #RP-98554).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.