Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLR3G Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546170
Description
Peptide sequence: SKRYMKVYKE EWIPDWRRLP REMMPRNKCK KAGPKPKKAK DAGKGTPLTN Sequence homology: Human: 100%.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3G is a specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It may direct with other members of the RPC3/POLR3C-RPC6/POLR3F-RPC7/POLR3G subcomplex RNA Pol III binding to the TFIIIB-DNA complex via the interactions between TFIIIB and POLR3F. It may be involved either in the recruitment and stabilization of the subcomplex within RNA polymerase III, or in stimulating catalytic functions of other subunits during initiation. POLR3G plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. POLR3G acts as nuclear and cytosolic DNA sensor involved in innate immune response. POLR3G can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.
Specifications
| POLR3G | |
| Polyclonal | |
| Unconjugated | |
| POLR3G | |
| 2310047G20Rik; AV275904; DNA-directed RNA polymerase III 32 kDa polypeptide; DNA-directed RNA polymerase III subunit G; DNA-directed RNA polymerase III subunit RPC7; Polr3g; polymerase (RNA) III (DNA directed) polypeptide G; polymerase (RNA) III (DNA directed) polypeptide G (32kD); polymerase (RNA) III subunit G; RNA polymerase III 32 kDa apha subunit; RNA polymerase III 32 kDa subunit; RNA polymerase III subunit C7; RNA polymerase III subunit G; RPC32; RPC32-alpha; RPC7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10622 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15318 | |
| POLR3G | |
| Synthetic peptide directed towards the middle region of human POLR3G. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction