Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3GL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | POLR3GL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17982620
![]() |
Novus Biologicals
NBP17982620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179826
![]() |
Novus Biologicals
NBP179826 |
100 μL |
Each for $499.50
|
|
|||||
Description
POLR3GL Polyclonal specifically detects POLR3GL in Human samples. It is validated for Western Blot.Specifications
POLR3GL | |
Polyclonal | |
Rabbit | |
NP_115681 | |
84265 | |
The immunogen for this antibody is POLR3GL. Peptide sequence DEEKEEEEEKEEEEEEEYDEEEHEEETDYIMSYFDNGEDFGGDSDDNMDE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
alternative RNA polymerase III subunit 32, DNA-directed RNA polymerase III subunit G-like, DNA-directed RNA polymerase III subunit RPC7-like, flj32422, FLJ34890, MGC3200, polymerase (RNA) III (DNA directed) polypeptide G (32kD) like, polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like, RNA polymerase III subunit C7-like, RPC32HOM, RPC32-like protein | |
POLR3GL | |
IgG | |
25 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title