Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3GL Antibody (OTI5E8), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273523G
Description
POLR3GL Monoclonal specifically detects POLR3GL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
POLR3GL | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POLR3GL | |
Full length recombinant protein of human POLR3GL (NP_115681) produced in HEK293T cells. | |
0.1 mL | |
84265.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5E8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
alternative RNA polymerase III subunit 32, DNA-directed RNA polymerase III subunit G-like, DNA-directed RNA polymerase III subunit RPC7-like, flj32422, FLJ34890, MGC3200, polymerase (RNA) III (DNA directed) polypeptide G (32kD) like, polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like, RNA polymerase III subunit C7-like, RPC32HOM, RPC32-like protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction