Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3H Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298184AF405
Description
POLR3H Polyclonal specifically detects POLR3H in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
POLR3H | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide, DNA-directed RNA polymerase III subunit H, DNA-directed RNA polymerase III subunit RPC8, KIAA1665MGC29654, MGC111097, polymerase (RNA) III (DNA directed) polypeptide H (22.9kD), RNA nucleotidyltransferase (DNA-directed), RNA polymerase III subunit 22.9 kDa subunit, RNA polymerase III subunit C8, RNA polymerase III subunit RPC8, RPC8RPC22.9 | |
Produced in rabbits immunized with E. coli-derived Human POLR3H fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
171568 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction