Learn More
Invitrogen™ POLR3H Monoclonal Antibody (OTI4D8)

Description
POLR3H Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | POLR3H |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | POLR3H |
| Gene Accession No. | Q9Y535 |
| Gene Alias | 5031409G22Rik; DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide; DNA-directed RNA polymerase III subunit H; DNA-directed RNA polymerase III subunit RPC8; KIAA1665; POLR3H; polymerase (RNA) III (DNA directed) polypeptide H; polymerase (RNA) III (DNA directed) polypeptide H (22.9kD); polymerase (RNA) III subunit H; RNA nucleotidyltransferase (DNA-directed); RNA polymerase III subunit 22.9 kDa subunit; RNA polymerase III subunit C7; RNA polymerase III subunit C8; RNA polymerase III subunit H; RPC22.9; RPC8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.