Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR3K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103798 Supplier Invitrogen™ Supplier No. PA5103798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPC10.

Eukaryotes produce 3 distinct classes of RNA polymerase, Pol I, II and III. Each polymerase is responsible for the synthesis of a different class of RNA. RNA polymerase III (Pol III) transcribes the 5S rRNA genes and all of the tRNA (transfer RNA) genes. POLR3K, also known as DNA-directed RNA polymerase III subunit RPC11, is a 108 amino acid nuclear protein that is one of the seventeen subunits that comprise Pol III. Specifically, POLR3K is a DNA-dependent RNA polymerase that uses the four ribonucleoside triphosphates to catalyze the transcription of DNA to RNA. The gene encoding POLR3K maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs and is associated with a variety of genetic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3K
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POLR3K
Gene Accession No. Q9CQZ7, Q9Y2Y1
Gene Alias 12.3kDa; 1500004O14Rik; AI196849; AU014893; C11; C11-RNP3; DNA-directed RNA polymerase III subunit K; DNA-directed RNA polymerase III subunit RPC10; DNA-directed RNA polymerases III 12.5 kDa polypeptide; estrous-specific protein, 250 kDa; hRPC11; HsC11p; My010; POLR3K; polymerase (RNA) III (DNA directed) polypeptide K; polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa; polymerase (RNA) III subunit K; RNA polymerase III 12.5 kDa subunit; RNA polymerase III subunit C10; RNA polymerase III subunit C11; RNA polymerase III subunit CII; RNA polymerase III subunit K; RPC10; RPC11; RPC12.5
Gene Symbols POLR3K
Host Species Rabbit
Immunogen A synthesized peptide derived from human POLR3K(Accession Q9Y2Y1), corresponding to amino acid residues G17-E67.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366277, 51728, 67005
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.