Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLRMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103801 Supplier Invitrogen™ Supplier No. PA5103801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total POLRMT.

This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLRMT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POLRMT
Gene Accession No. O00411
Gene Alias 1110018N15Rik; 4932416K13; APOLMT; DNA-directed RNA polymerase, mitochondrial; h-mtRPOL; LOW QUALITY PROTEIN: DNA-directed RNA polymerase, mitochondrial; mitochondrial DNA-directed RNA polymerase; MTRNAP; MTRPOL; POLRMT; polymerase (RNA) mitochondrial (DNA directed); RNA polymerase mitochondrial
Gene Symbols POLRMT
Host Species Rabbit
Immunogen A synthesized peptide derived from human POLRMT(Accession O00411), corresponding to amino acid residues S784-G834.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5442
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.