Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poltex Flex Neck Biohazard Shield with Magnetic Base

Click to view available options
Length (English):
8.5 in.
12 in.
Description
- Made from durable scratch and break-resistant polycarbonate
- Available in two sizes - 8.5 x 12 in. and 12 x 12 in.
Specifications
Specifications
| Dimensions (L x W) | 8.5 x 12 in. |
| Width (English) | 12 in. |
| Product Type | Flexible Neck Biohazard Shield |
| Includes | Magnetic base |
| Length (English) | 8.5 in. |
| Material | Polycarbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction