Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylamide), carboxyl modified, pure, low carboxyl content, ∽ MW 200,000, Thermo Scientific™

Catalog No. p-185651
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 62649-23-4
Molecular Formula (C3H5NO)n
Molecular Weight (g/mol) 237.19
MDL Number MFCD00084393
InChI Key DTBMFUOWAASNDB-UHFFFAOYSA-M
Synonym 2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate, sodium acrylamide acrylic acid acrylate
PubChem CID 23696299
IUPAC Name sodium prop-2-enamide prop-2-enoic acid prop-2-enoate
SMILES NC(=O)C(-*)C-*

Specifications

Color White
Physical Form Powder or Granules
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol
Quantity 5 g
Formula Weight ≈200.000
Grade Pure
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Chemical Name or Material Poly(acrylamide), carboxyl modified, Carboxyl Modified, Low Carboxyl Conten

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.