Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000

Catalog No. AC178042500
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle

CAS: 9003-05-8 | (C3H5NO)n | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-05-8
Molecular Formula (C3H5NO)n
MDL Number MFCD00084392
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C(-*)C-*

Specifications

Color White to Yellow
Physical Form Granules
Linear Formula [-CH2CH(CONH2)-]n
Density 1.3000g/mL
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid
Quantity 250 g
IUPAC Name prop-2-enoic acid
Molecular Weight (g/mol) 72.06
PubChem CID 6581
ChEBI CHEBI:18308
Formula Weight 5 to 6.000.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 13% max.
Packaging Glass bottle
Specific Gravity 1.3
Chemical Name or Material Poly(acrylamide)
Show More Show Less
RTECSNumber AS3700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.