Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-D-Lysine Hydrobromide, MP Biomedicals™

Catalog No. mp215017580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215017580 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215017580 Supplier MP Biomedicals, Inc Supplier No. 0215017580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhances cell adhesion to solid substrates. Poly-D-Lysine Hydrobromide, MP Biomedicals is a synthetic amino acid that binds to DNA, red cell membrane and any negatively charged protein. It has been shown to eliminate prion proteins from infected cells. Mo

When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding. It is a compromise between easier-to-use lower molecular weight products and the extremely viscous higher molecular weights.

Poly-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. Both the D- and L- forms can be used as a coating substrate, since poly-lysine is a nonspecific attachment factor for cells; however, certain cells can digest it. In this case, poly-D-lysine should be used as the attachment factor so that the cells are not disrupted by excessive uptake of L-lysine.

Poly-D-Lysine Hydrobromide is typically used as a coating substrate for culture dishes, slides, etc. Other uses for poly-lysine have been reported as well:

  • Conjugation to methotrexate for increased drug transport
  • Microencapsulation of islets
  • Use in simple reproducible procedure for chromosomal preparations from a variety of tissues
  • Immobilation of living cells in biocompatible semipermeable microcapsules
  • In the preparation of polycationic beads
  • Conjugation to albumin and horseradish peroxidase to enhance cellular uptake
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27964-99-4
Molecular Formula C6H15BrN2O2
Molecular Weight (g/mol) 227.102
InChI Key MEXAGTSTSPYCEP-NUBCRITNSA-N
Synonym d-lysine hydrobromide
PubChem CID 87493163
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrobromide
SMILES C(CCN)CC(C(=O)O)N.Br

Specifications

Solubility Information 50mg/mL, H2O
Quantity 100 mg
Formula Weight 70000-150000
Physical Form Powder
Chemical Name or Material Poly-D-lysine Hydrobromide
Recommended Storage Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.