Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol), average M.W. 1500, ACROS Organics™

Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
25322-68-3
MFCD01779609
PEG
25322-68-3
MFCD01779609
PEG
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Hydroxyl Value 70 to 80mg KOH/g
Formula Weight 1500
Physical Form Waxy Flakes
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: 650g/L (20°C). Other solubilities: soluble in many organic solvents
Water 0.5% max. (K.F.)
Viscosity 36-42 mPa.s (20°C)
Color White
Melting Point 44.0°C to 48.0°C
Quantity 100 g
Flash Point >250°C
Chemical Name or Material Poly(ethylene glycol), Average M.W. 1500
Show More Show Less

RTECSNumber : TQ4100000

TSCA : TSCA

RMF - Research Use or Further Mfg