Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium acetate tetrahydrate, 96%

Catalog No. AC194941000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Phase transfer catalyst | CAS: 67533-12-4 | C10H23NO2·4H2O | 261.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67533-12-4
Molecular Formula C10H23NO2·4H2O
Molecular Weight (g/mol) 261.35
InChI Key QNOAKQGNSWGYOE-UHFFFAOYSA-M
Synonym tetraethylammonium acetate tetrahydrate, ethanaminium, n,n,n-triethyl-, acetate, tetrahydrate, tetraethylazanium acetate tetrahydrate, tetraethylazanium ethanoate tetrahydrate, acetate ion tetraethylammonium tetrahydrate, ethanaminium,n,n,n-triethyl-,acetate,tetrahydrate
PubChem CID 2724098
IUPAC Name tetraethylazanium;acetate;tetrahydrate
SMILES CC[N+](CC)(CC)CC.CC(=O)[O-].O.O.O.O

Specifications

Melting Point 40.0°C to 45.0°C
Color Colorless to White
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (C2H5)4N(O2CCH3)·4H2O
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 261.35
Percent Purity 97+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium acetate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.