Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol), average M.W. 6000

Catalog No. p-4599881
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC192280010

Supplier: Thermo Scientific Chemicals 192280010

Only null left
Add to Cart

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form White flakes or powder
Melting Point 55.0°C to 60.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in thf
Quantity 1 kg
Formula Weight 6000
Hydroxyl Value 17 to 20mg KOH/g
Packaging Plastic bottle
Viscosity 220-262 mPa.s (20°C)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less
EINECSNumber 500-038-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.