Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly(ethyleneimine) Solution, 50% (w/v) in Water, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
100 mL
500 mL
1 L
Specifications
Specifications
CAS | 9002-98-6 |
Concentration | 50% (w/v) in H 2 O |
Synonym | Ethyleneimine polymer solution; PEI |
Type | Analytical Standard |
Quantity | 500 mL |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction