Learn More
Description
Poly(I:C) is a synthetic double-stranded RNA (dsRNA); endosomal Toll-like receptor 3 (TLR3) agonist. It also binds to the cytoplasmic receptors, retinoic acid-inducible gene I (RIG-I) and melanoma differentiation-associated gene 5 (MDA-5), elicits the secretion of type I interferon (IFN) and pro-inflammatory cytokines. Poly(I:C) is commonly used for immune system activation in vivo. Poly(I:C) also promotes maturation of hPSC-derived cardiomyocytes by modulating early Notch signaling and cardiomyogenic transcriptional programs. Poly(I:C) promotes generation of stable, mature dendritic cells in combination with TLR8 agonist CL 075 . Transfection into NIT-1 β cells induce β-cell apoptosis. Commonly used to stimulate immune system activation in vivo. Also promotes maturation of hPSC-derived cardiomyocytes.
To prevent denaturation, this product should be reconstituted in solutions with physiological salt concentrations.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Purity | >98% |
| Molecular Weight (g/mol) | 671.406 |
| Chemical Name or Material | Poly(I:C) |
Safety and Handling
| Recommended Storage | Store at -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

