Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals

Catalog No. p-2625542 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 mg
100 mg
500 mg

Chemical Identifiers

CAS 25988-63-0
Molecular Formula C18H38N6O4
Molecular Weight (g/mol) 402.54
InChI Key WBSCNDJQPKSPII-KKUMJFAQSA-N
Synonym lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
PubChem CID 72363
IUPAC Name (2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
SMILES C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N

Specifications

Quantity 25 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-L-Lysine Hydrobromide
Recommended Storage 0°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.