Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

poly-(LPAC) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN225810MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN225810MI 100 μL
ABN225825MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN225810MI Supplier MilliporeSigma Supplier No. ABN2258100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

poly-(LPAC) Polyclonal antibody specifically detects poly-(LPAC) in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen poly-(LPAC)
Applications Immunohistochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias (Leu-Pro-Ala-Cys)Exp
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the (LPAC)EXP repeat motif.
Molecular Weight of Antigen ∼25 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects proteins with poly-(LPAC).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.