Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(methyl methacrylate), beads, average M.W. 35,000

Catalog No. p-186389
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
Synonym Methyl methacrylate, polymerized, PMMA
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Color White
Physical Form Beads
Melting Point 105.0°C
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Density 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 25 g
Flash Point 300°C
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Particle Size Bead size (typically): 50 to 150 micron
Refractive Index 1.49
Chemical Name or Material Poly(methyl methacrylate), nominal M.W. 38000
Show More Show Less
RTECSNumber TR0400000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.