Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230

Catalog No. p-7052598
Click to view available options
Quantity:
100 mL
2.5 L

Catalog No. AAH37543AE

Supplier: Thermo Scientific Chemicals H37543AE

Only null left
Add to Cart

CAS: 9046-10-0 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9046-10-0
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
InChI Key JCZPOYAMKJFOLA-UHFFFAOYNA-N
Synonym 3,4-pyrrolidinediol, 3,4-dihydroxy-2h-pyrroline, polyetheramine d 230, polyetheramine d 2000, poly propylene glycol bis 2-aminopropyl ether, poly propylene oxide , diamine terminated, o,o'-bis 2-aminopropyl polypropyleneglycol, o,o'-bis 2-aminopropyl polypropylene glycol 300, polypropylene glycol 130 bis 2-aminopropyl ether, polypropylene glycol 300 bis 2-aminopropyl ether
PubChem CID 13329881
IUPAC Name pyrrolidine-3,4-diol
SMILES OC1CNCC1O

Specifications

Color Colorless to Yellow
Density 0.948 g/mL
Quantity 100 mL
UN Number UN2922
Physical Form Liquid
Chemical Name or Material Poly(propylene glycol) bis(2-aminopropyl ether), Mn ≈230
Hazard Category H301+H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECSNumber TR3702500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.