Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab MasterTech Special Stain Kit, Potassium Ferrocyanide

Click to view available options
:
500 g
Description
- A MasterTech Special Stain Kit Component
- Used in the Prussian blue staining technique: Detecting iron deposits in tissues by reacting with ferric ions to form a blue precipitate known as Prussian blue
- Also used for identifying iron deficiency or excess iron: Used for identifying anemia or hemochromatosis.
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Color | Yellow |
| Quantity | 500 g |
| Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction