Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(sodium-p-styrenesulfonate), average M.W. 70.000

Catalog No. p-186478
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Color Tan to White
Physical Form Powder
Linear Formula [-CH2CH(C6H4SO3Na)-]n
Solubility Information Solubility in water: soluble. Other solubilities: soluble in lower glycerols
Quantity 5 g
IUPAC Name 4-ethenylbenzene-1-sulfonic acid
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Poly(sodium-p-styrenesulfonate), Average M.W. 70.000
RTECSNumber DB8550000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.