Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number AA4265322
Click to view available options
Quantity:
100 g
500 g
28210-41-5
MFCD00165973
4-styrenesulfonic acid, polystyrene sulfonate, tolevamer, styrene-4-sulphonic acid, 4-vinylbenzenesulfonic acid, benzenesulfonic acid, 4-ethenyl, poly 4-vinylbenzenesulfonic acid, tolevamer inn, poly styrene sulfonic acid sodium salt, unii-zsl2fb6gxn
28210-41-5
MFCD00165973
4-styrenesulfonic acid, polystyrene sulfonate, tolevamer, styrene-4-sulphonic acid, 4-vinylbenzenesulfonic acid, benzenesulfonic acid, 4-ethenyl, poly 4-vinylbenzenesulfonic acid, tolevamer inn, poly styrene sulfonic acid sodium salt, unii-zsl2fb6gxn

Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Amber
Density 1.1 g/mL
Quantity 100 g
UN Number UN2584
Solubility Information Miscible with water,ethanol,methanol,lower alcohols and glycols.
Concentration or Composition (by Analyte or Components) 30% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Poly (styrene sulfonic acid), M.W. 75,000
Hazard Category H314-H335-H350
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit