Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL401 Polyurethane Coated Nylon Gloves

Catalog No. p-7182065 Shop All Pyramex Safety Products Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are designed to enhance grip capabilities without being sticky, in addition to providing a high degree of dexterity and tactile sensitivity.

  • 13 gauge nylon gloves with polyurethane coating
  • Provides dexterity and tactile sensitivity
  • Enhanced grip capabilities
  • Low particulate shed

Specifications

ANSI Abrasion Rating ANSI 1
Cuff Material Latex Elastic
Coating Material Polyurethane
Color Gray
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 4131X
For Use With (Application) Construction, Electronics, Inspection, and Quality Control
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series GL401
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.