Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydrogen hexachloroplatinate(IV) hydrate, ACS reagent

Catalog No. AC405010050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid hydrate, Platinic chloride hydrate
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60.0°C
Physical Form Chunks and/or Powder
Quantity 5 g
Linear Formula H2PtCl6·xH2O
Solubility Information (in alcohol) Passes test
Formula Weight 409.82
Percent Purity ≥37.50% (Pt)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Hydrogen hexachloroplatinate(IV) hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.