Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Polybrene
GREENER_CHOICESDP

Catalog No. 771110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-111-0 Supplier R&D Systems™ Supplier No. 7711/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Viral transduction enhancer

Polybrene is a cationic polymer that improves lentiviral and adenoviral transduction efficiency in mammalian cells in vitro. It also enhances DNA transfection in many types of cells. It has a neutralizing effect on negatively charged DNA and viral particles which facilitates their attachment and entry into host cells. Polybrene is suitable for use with CRISPR/Cas9 protocols.

Polybrene is a lyophilized solid and can be prepared as a stock solution by adding 1 mL of sterile ultra-pure water directly to the vial (final stock concentration 10 mg/mL).

For further information view the product protocol .

Specifications

Chemical Name or Material 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide
CAS 28728-55-4
Quantity 10 mg
Product Line Tocris
Target Viral Transduction Enhancers
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.