Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Polybutadiene, cis
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
Description
Polybutadiene, cis (PB-cis) is a synthetic rubber that is prepared by solution polymerization of 1,4-butadiene. Its mechanical properties include low temperature flexibility and abrasion resistance. It can be cured by using sulfur based donors and peroxides and thus PB-cis acts as a highly cross-linked polymer with high resilience.PB-cis can be polymerized with styrene for the formation of high impact polystyrene (HIPS). It may also be used as a toughening agent for acrylonitrile butadiene rubber (ABS) based resins.
Specifications
Specifications
| CAS | 9003-17-2 |
| Density | 0.9 g/mL (at 25°C) |
| Linear Formula | (CH2CH=CHCH2)n |
| Quantity | 100 g |
| MDL Number | MFCD00084398 |
| Molecular Weight (g/mol) | 200,000-300,000 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction