Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Protective Boot Covers

Catalog No. p-4246893
Click to view available options
Color:
Blue
Yellow
Size:
Large
Regular
Thickness:
4 mil
6 mil
This item is not returnable. View return policy
This item is not returnable. View return policy

Provide knee-high, foot and lower leg protection in situations where biological and chemical liquids are present

  • Knee-high cover with elastic top
  • Two supple weights provide flexible, lightweight coverage
  • Available in different colors
  • Standard or cleanroom packaged (for Class 100 controlled environments)

Specifications

Size Regular
Material Polyolefin
Color Blue
Closure Type Elastic Top
Seam Type Heat sealed
Thickness 4 mil
For Use With (Application) Ideal for use where biological and chemical liquids are present
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.