Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Protective Sleeves

Catalog No. 19125504
Click to view available options
Thickness:
4 mil
6 mil
Size:
Standard
This item is not returnable. View return policy

Catalog No. 19-125-504

Supplier: PolyCo™ 41803

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Easy to clean and reuse, VR sleeves are lightweight yet more durable than vinyl.

  • Waterproof - barrier protection
  • AAMI Class 2 chemical splash protection
  • Ideal for flexibility
  • Strong, natural rubber elastic cuffs
  • Textured inside surface minimizes cling and enhances ventilation
  • Smooth outside surface for ease of cleaning
  • Available in different colors and thicknesses
  • Made in USA

 

Specifications

Size Standard
Material VR™ Polyolefin
Disposable Reusable
Cuff Style Elastic
Finish Textured
Model 41803
Thickness 6 mil
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Certifications/Compliance 21 CFR 175.300
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.