Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polycystin-1/PKD1 Antibody (PKS161f/B7), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250247AF488
Description
Polycystin-1/PKD1 Monoclonal specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Polycystin-1/PKD1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PKD1 | |
| Recombinant protein (233 aa) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| PKS161f/B7 | |
| Western Blot, Immunohistochemistry | |
| Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5310 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction