Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polycystin-1/PKD1 Antibody (PKS161f/B7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Polycystin-1/PKD1 |
---|---|
Clone | PKS161f/B7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry |
Description
Polycystin-1/PKD1 Monoclonal antibody specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Polycystin-1/PKD1 | |
1 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Phospho Specific | |
PBS | |
5310 | |
IgG1 κ | |
Protein A purified |
PKS161f/B7 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1 | |
Recombinant protein (233 aa) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title