Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Polycystin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142387
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142387 Supplier Invitrogen™ Supplier No. PA5142387
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_001009944.1 and NP_000287.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Involved in renal tubulogenesis. Involved in fluid-flow mechanosensation by the primary cilium in renal epithelium. Acts as a regulator of cilium length, together with PKD2. The dynamic control of cilium length is essential in the regulation of mechanotransductive signaling. The cilium length response creates a negative feedback loop whereby fluid shear-mediated deflection of the primary cilium, which decreases intracellular cAMP, leads to cilium shortening and thus decreases flow-induced signaling. May be an ion-channel regulator. Involved in adhesive protein-protein and protein-carbohydrate interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polycystin 1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Pkd1
Gene Accession No. P98161
Gene Alias Autosomal dominant polycystic kidney disease 1 protein; autosomal dominant polycystic kidney disease 1 protein homolog; mFLJ00285; PBP; PC1; Pc-1; PKD1; polycystic kidney disease 1 (autosomal dominant); polycystic kidney disease 1 homolog; polycystic kidney disease 1 homolog; polycystin-1; polycystic kidney disease-associated protein; polycystin 1; polycystin 1, transient receptor potential channel interacting; polycystin-1; transient receptor potential cation channel, subfamily P, member 1; TRPP1
Gene Symbols Pkd1
Host Species Goat
Immunogen Peptide with sequence C-RTPLRAKNKVHP.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.