Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polycystin 2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130117AF532
Description
Polycystin 2 Polyclonal antibody specifically detects Polycystin 2 in Human, Mouse samples. It is validated for Western BlotSpecifications
Polycystin 2 | |
Polyclonal | |
Western Blot | |
APKD2, Autosomal dominant polycystic kidney disease type II protein, MGC138466, MGC138468, Pc-2, PC2, PKD4, polycystic kidney disease 2 (autosomal dominant), Polycystic kidney disease 2 protein, polycystin-2, Polycystwin, R48321, transient receptor potential cation channel, subfamily P, member 2, TRPP2 | |
maps to a region between residue 918 and 968 of human polycystin-2 using the numbering given in entry NP_000288.1 | |
0.1 mL | |
Cancer, Protein Kinase, Signal Transduction, Tumor Suppressors | |
5311 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction