Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polycystin 2 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306363UV
Description
Polycystin 2 Polyclonal specifically detects Polycystin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Polycystin 2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
APKD2, Autosomal dominant polycystic kidney disease type II protein, MGC138466, MGC138468, Pc-2, PC2, PKD4, polycystic kidney disease 2 (autosomal dominant), Polycystic kidney disease 2 protein, polycystin-2, Polycystwin, R48321, transient receptor potential cation channel, subfamily P, member 2, TRPP2 | |
Produced in rabbits immunized with E. coli-derived Human Polycystin 2 fragment. | |
0.1 mL | |
Cancer, Protein Kinase, Signal Transduction, Tumor Suppressors | |
5311 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction