Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polycystin 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984549
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4549 Supplier Bioss Supplier No. BS2158R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal dominant polycystic kidney disease.

Specifications

Antigen Polycystin 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PKD2
Gene Accession No. Q13563
Gene Alias A MGC138466; APKD2; Autosomal dominant polycystic kidney disease type II protein; C030034P18Rik; MGC138468; PC2; Pc-2; Pkd2; PKD4; polycystic kidney disease 2; polycystic kidney disease 2 (autosomal dominant); polycystic kidney disease 2 homolog; Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; polycystin 2; polycystin 2, transient receptor potential cation channel; Polycystin-2; Polycystwin; R48321; RGD1559992; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2; TRPP1; TRPP2
Gene Symbols PKD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Polycystin 2 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5311
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.