Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250

Manufacturer:  Alfa Aesar™ 42498K2

Quantity 1L

 View more versions of this product

Catalog No. AA42498K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

9016-00-6
MFCD00148360
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1,250

Specifications

9016-00-6
Liquid
Odorless
>205°C
−40°C
263°C (505°F)
0.935g/mL
MFCD00148360
1.3990
Colorless
1L
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1,250

Safety and Handling

SDS & Safety Documents

RTECSNumber : TQ2690000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit