Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4000
Description
Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Physical Form | Viscous Liquid |
Boiling Point | >225°C |
Melting Point | <-40°C |
Density | 0.960 g/mL |
Solubility Information | Slightly miscible with water. Immiscible with methanol and ethylene glycol. |
Quantity | 250 mL |
Flash Point | 285°C (545°F) |
Odor | Odorless |
Refractive Index | 1.4015 |
Show More |
Safety and Handling
RTECSNumber | TQ2690000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.