Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000

Catalog No. AA44764K2
Click to view available options
Quantity:
50 mL
250 mL
1 L
4 L

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppresses the formation of foams and is a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Physical Form Liquid
Density 0.974 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 1 L
Odor Odorless
Refractive Index 1.4035
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.