Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyester acrylate/methacrylate, PEAM-645

Catalog No. p-7080158
Click to view available options
Quantity:
25 g
100 g

Recommended for use as a base resin in adhesive applications or coating applications | CAS: 921213-39-0

Recommended for use as a base resin in adhesive applications or coating applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921213-39-0

Specifications

Color Amber to Brown
Physical Form Liquid
Melting Point >375°C (decomposition)
Quantity 25 g
Formula Weight 645
Odor Mild
Chemical Name or Material Polyester acrylate/methacrylate, PEAM-645
Hazard Category H304-H315-H317-H318-H335-H336-H361
Hazard Statement GHS H Statement
H361-H315-H319-H317-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P331-P333+P313-P362-P501c
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.