Alfa Aesar™ Polyester acrylate/methacrylate, PEAM-645

Recommended for use as a base resin in adhesive applications or coating applications

Manufacturer: Alfa Aesar™ 4659614

Quantity 25g

 View more versions of this product

Catalog No. AA4659614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 921213-39-0
    Formula Weight 645
    Chemical Name or Material Polyester acrylate/methacrylate, PEAM-645

    Specifications

    Formula Weight 645
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H361-H315-H319-H317-H335
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    Physical Form Liquid
    CAS 921213-39-0
    Health Hazard 1 Warning
    Odor Mild
    Color Dark amber to brown
    Melting Point >375°C (dec.)
    Quantity 25g
    Chemical Name or Material Polyester acrylate/methacrylate, PEAM-645

    GHS H Statement
    H361-H315-H319-H317-H335
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit