Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol 1,500

Catalog No. p-7023145
Click to view available options
Quantity:
250 g
1000 g

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Cell fusion induced by polyethylene glycol(PEG) has become a standard method in somatic cell genetics. PEG promoted cell fusion is involved in the production of hybridoma cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 1500
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point 44°C to 48°C
Merck Index 14,7568
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Quantity 250 g
Odor Odorless
Chemical Name or Material Polyethylene glycol 1,500
EINECSNumber 500-038-2
RTECSNumber TQ4030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.