Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 1000, NF, Spectrum™ Chemical
SDP

Catalog No. 18605299
Click to view available options
Quantity:
2.5 kg
6 x 500 g
Packaging:
Amber Glass Bottle
Poly Bottle

PO118, 25322-68-3, H(OCH2CH2)nOH

Spectrum™ Chemical Polyethylene Glycol 1000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 4.5 to 7.5
CAS Max % 1
Quantity 2.5 kg
Grade NF
Ignition Residue 0.001
Packaging Poly Bottle
Viscosity 16.0-19.0 cSt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.