Learn More
Polyethylene glycol 200
Description
Polyethylene glycol is used in electron microscopy. It is used as a solvent, a dye carrier in inks and a dispersant in water based coatings. Its derivatives like PEG esters are used as surfactants. In the textile industry, it is used as a lubricant, softener, dye carrier dispersant and an anti-static agent. It finds application in personal care products like skin creams, lotions and bath gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Liquid |
Melting Point | °C to 55 to°C to 40°C |
Density | 1.127 |
Merck Index | 14,7568 |
Solubility Information | Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons. |
Quantity | 1000 g |
Formula Weight | ≈200 |
Flash Point | 171°C (339°F) |
Refractive Index | 1.459 |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H227-H303-H315-H320-H333-H335 |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c |
EINECSNumber | 203-473-3 |
RTECSNumber | TQ3600000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.